JACKSON SAFETY™ V80 SG34™ Protective Goggles, KIMBERLY-CLARK PROFESSIONALl®
Supplier: Ansell Healthcare
16362
89215-604CS 509.72 USD
89215-604
JACKSON SAFETY™ V80 SG34™ Protective Goggles, KIMBERLY-CLARK PROFESSIONALl®
Goggles
KleenGuard™ SG34 goggle with a soft, pliable frame for enhanced fit and comfort with button vents provide indirect venting for comfort.
  • Economical splash protection
  • 50 KleenGuard™ V80 SG34 safety goggles/case; clear lens with green frame; uiversal size (one size fits most)
This goggle fits over most prescription eyeglasses and the headband buckles are located toward the rear of the frame to limit interference with peripheral vision on the job. The clear lenses (with green frame) allow maximum visible light transmission and are ideal for indoor use.More Product Information