45/50 Large Scale Moisture Test Receiver, Barrett Type, Ace Glass Incorporated
Supplier: Ace Glass
7744-50
89215-158EA 4182.58 USD
89215-158
45/50 Large Scale Moisture Test Receiver, Barrett Type, Ace Glass Incorporated
Distillation Components Distillation Receivers
One liter Barrett type moisture test receiver
2 mm PTFE stopcock on bottom for draining contents. Standard taper 45/50 joints at top and on side arm. 60 mm distance between side arm and body for clearance on spherical or cylindrical reactor bodies. Graduated in 10 mL subdivisions. Fabricated from boroilicate glass.