Duran Bottle GL 80 Connection System, Ace Glass Incorporated
Supplier: Ace Glass
7631-04 7631-03 7631-02 7631-01 7631-13 7631-14 7631-10 7631-11 7631-12
89214-906EA 116.12 USD
89214-906 89214-908 89214-910 89214-928 89214-932 89214-934 89214-938 89214-942 89214-946
Duran Bottle GL 80 Connection System, Ace Glass Incorporated
Closures Bottle Closures
The heart of the GL 80 connection system is a GL80 polypropylene cap with 4, GL18 threaded ports
Modular accessories include polypropylene tube connection caps,solid polybutylene terepthalate (PBT) caps, PBT pressure equalization caps and PTFE cap inserts to accommodate various sizes of tubing. A typical application is the safe transfer of liquid media within a closed and sterile system for which you'd need one two-pack code 89214-928 GL80 cap, two 89214-906 equalization caps, and some combination of solid caps and tubing connections with inserts