Greenbox™ Shipping Box Pre-Qualified Solutions Shipping System, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
09UF2 09UR1 09UR2 09UC3 09UC5 12UF1 12UF3 12UR1 12UR2 12UR3 12UR4 12UC4 12UC5 24UF4 24UR3 24UC6
89213-020EA 282.02 USD
89213-020 89213-022 89213-024 89213-026 89213-028 89213-030 89213-032 89213-034 89213-036 89213-038 89213-040 89213-042 89213-044 89213-046 89213-048 89213-050
Greenbox™ Shipping Box Pre-Qualified Solutions Shipping System, Sonoco ThermoSafe
Mailers and Shippers
Greenbox Reusable Insulated Shippers have been designed to utilize simple packaging configurations, reduce operational complexity, and optimize total cost of ownership.
  • No waiting
  • Fewer components and lower weight
  • Biodegradable
  • Simple design
Ensure the efficacy of your valuable temperature sensitive products and maintain regulatory compliance throughout distribution with Greenbox.
Zero Bench-Time™ refrigerant design means no waiting, greatly simplifies and accelerates the assembly process. Utilizes VIP insulation and PureTemp® PCMs resulting inMore Product Information