MICRO-THERM USB Temperature Logger
Supplier: Thermco
ACCD00408
89206-464EA 240.17 USD
89206-464
MICRO-THERM USB Temperature Logger
Data Loggers
Small, rugged, waterproof data logger for monitoring and recording temperatures from –40 to 80°C (–40 to 176°F)
Data can be viewed on the three-digit LCD or downloaded via the USB connector. Allows for sample rates of one per second to one per two hours, and stores up to 8000 samples.Visual alarm indicates when the temperature crosses a predefined threshold. Battery is easily replaceable. Includes MICROLab Lite software and manual