PIG® Traffic Mat® Heavy-Duty Absorbent Rug, New Pig
Supplier: New Pig
MAT162
89205-916EA 363.56 USD
89205-916
PIG® Traffic Mat® Heavy-Duty Absorbent Rug, New Pig
Spill Control Materials
100% polypropylene material is heat fused and needle punched for incredible durability for walk-on and wheeled traffic
  • Absorbs and retains oils, coolants, solvents, and water
  • Dark gray color hides grime, so mat stays on the job longer
  • Rolls are ideal for walkways and large areas to soak up leaks and overspray
  • Cuts easily with scissors or utility knife
  • Can be incinerated after use to reduce waste or for fuels blending
  • Heavy-duty rug is twice as heavy and absorbent as standard traffic mat rug
Our most durable mat won't rip, shred or fray, even under heavy forklift traffic.