SGE Syringes, eVol® XR Digital Analytical Syringe, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
2910020 2910022 2910024 2910021 2910023 2910025 2910026 2910027 2910320 2910322 2910324 2910035 2910037 2910335 2910329 2910029
89194-536EA 2261.55 USD
89194-536 97049-424 97049-426 97049-428 89205-470 89205-472 89205-474 89205-476 89205-478 89205-480 89205-482 89205-484 89236-486 89236-488 89236-490 89236-494 89368-640 89368-642
SGE Syringes, eVol® XR Digital Analytical Syringe, Trajan Scientific and Medical
Syringes Syringes for Chromatography
eVol® XR is the coupling of two precision devices: a digitally controlled electronic drive and an XCHANGE® enabled analytical syringe.
  • Ergonomic, comfortable and easy to use
  • Variable speed of operation
  • Easily and quickly changed to dedicate to individual liquids or methods
  • Easily calibrated by operators
  • Inject directly onto a chromatography column with a consistent flow rate
The device is easily calibrated by operators, and calibration factors saved for each syringe, enabling laboratories to comply with stringent global laboratory standards (e.g. GLP,More Product Information