PURELAB® Classic Water Purification Systems, ELGA LabWater
Supplier: ELGA LabWater
CLXXXUFM2-US CLXXXDIM2-US CLXXUVFM2-US CLXXXUVM2-US SK-CSPCLASSICA SK-CSPCLASSICB SKCSPCLASSICUV
89204-080EA 7301.66 USD
89204-080 89204-076 89204-072 89204-084 76516-392 76516-394 76516-396
PURELAB® Classic Water Purification Systems, ELGA LabWater
Water Purification Systems Pure and Ultra Pure Water Systems
The PURELAB Classic system combines high performance with economy to deliver 18.2 MΩ-cm type I ultrapure water at a very cost-effective price
  • Ultrapure type I water at very economic costs for the equipment and the cost of ownership
  • Automatic intermittent recirculation minimizes temperature build-up and optimizes microbial performance
  • Very easy to maintain – incorporates ‘fast rinse’ ultra filter
  • Upgradable from single pack to twin pack purification
  • Suitable for all life science and analytical applications
  • Complete sanitization of all wetted parts ensures
More Product Information