ECO Air-Cooled Refrigerated Circulators, Lauda-Brinkmann
Supplier: Lauda-Brinkmann
L001433 L001434 L001438 L001439 L001437 L001440 L001441 L001443 L001445 L001446 L001444 L001405 L001408 L001410 L001411 L001409 L001412 L001415 L001417 L001418 L001416
89202-386EA 8317.81 USD
89202-386 89202-388 89202-390 89202-394 89202-396 89202-398 89202-400 89202-402 89202-404 89202-406 89202-408 89202-410 89202-412 89202-478 89202-480 89202-482 89202-484 89202-486 89202-488 89202-490 89202-492 89202-494 89202-496 89202-498 89202-500 89202-502 89202-504
ECO Air-Cooled Refrigerated Circulators, Lauda-Brinkmann
Circulating Thermostats Closed Circulator Systems Cooling Circulators
ECO current replacement for Ecoline
The intuitive menu navigation allows for easy operation. Both control heads are equipped with a mini-USB interface as a standard feature. Additional interfaces are available via the add-on modules. On-the-fly adjustment of the flow between internal and external circulation is made possible even during operation via the new flow rate switch. The more powerful units are equipped with the energy-saving LAUDA SmartCool system. All cooling thermostats are available with air-cooled or water-cooled options. Stainless steel bath.
There are innovations andMore Product Information