ECO Heating Circulators with Transparent Bath, Lauda-Brinkmann
Supplier: Lauda-Brinkmann
L001203 L001204 L001205 L001206 L001207 L001210 L001183 L001184
89202-346EA 5808.08 USD
89202-346 89202-348 89202-350 89202-352 89202-354 89202-356 89202-358 89202-360 89202-438 89202-440 89202-442 89202-444 89202-446 89202-448 89202-450 89202-452
ECO Heating Circulators with Transparent Bath, Lauda-Brinkmann
Circulating Thermostats Closed Circulator Systems Heating Circulators
ECO current replacement for Ecoline
The intuitive menu navigation allows for easy operation. Both control heads are equipped with a mini-USB interface as a standard feature. Additional interfaces are available via the add-on modules. On-the-fly adjustment of the flow between internal and external circulation is made possible even during operation via the new flow rate switch. The more powerful units are equipped with the energy-saving LAUDA SmartCool system. All cooling thermostats are available with air-cooled or water-cooled options. Transparent bath.
There are innovations andMore Product Information