Simport SnapTwist™ Micrewtube®
Supplier: Simport Scientific
T343BLS T343LLS T343LLSL T343GLSL T342-4T T342-5T T342-6T T342-7T T343YLS T343WLS T343NLSL T343RLSL T343YLSL T343NLS T343RLS T343GLS T343BLSL T343WLSL
89202-000PK 140.81 USD
89202-000 89202-004 89202-018 89202-016 89201-990 89201-992 89201-994 89201-996 89202-008 89202-010 89202-012 89202-020 89202-022 89201-998 89202-006 89202-002 89202-014 89202-024
Simport SnapTwist™ Micrewtube®
Tubes Microtubes
The SnapTwist Micrew Provides all the advantages of a modern microcentrifuge tube with screw cap but the SnapTwist closure is a true time saver.
  • Conical bottom tubes can be centrifuged up to 20000 g
  • Temperature range: -90 to 100 °C
  • Certified RNase-, DNase-, Pyrogen- and DNA-free
  • Tube made of polypropylene
  • Cap made of polyethylene
The SnapTwist Micrew has a multitude of applications around your lab. It is ideal for freezer storage, boiling application, centrifugation and more, and will fit most standard microcentrifuge rotors.
The tubes can beMore Product Information