Cap, GL Thread, PTFE Membrane, Ace Glass Incorporated
Supplier: Ace Glass
7629-25 7629-32 7629-45 7629-80
89199-114CS 342.16 USD
89199-114 89199-116 89199-118 89199-120
Cap, GL Thread, PTFE Membrane, Ace Glass Incorporated
Closures Bottle Closures
These polypropylene, blue caps have a center hole and welded-in, PTFE, inner gas permeable membrane
Ideal for media sterilization and storage. The Temperature Range (°C) for all modules is -40 to + 140°C.