IHC Control Slides, Azer Scientific
Supplier: VWR International
89169-026 89169-030 89169-032 89169-034 89169-040 89169-042
89169-026PK 340.24 USD
89169-026 89169-030 89169-032 89169-034 89169-040 89169-042
IHC Control Slides, Azer Scientific
Slides Microscopy Control Slides
These control slides are designed specifically for quality control of immunohistochemistry staining protocols.
  • Ten unstained slides and one stained slide per box
  • PIN-4 slides: 20 negative slides per box
  • For quality control of immunohistochemistry staining protocols
Each set contains 10 unstained slides and one stained slide, except the PIN-4 slides, which have 20 negative slides per box.
IHC Control Slides contain specific characteristics that are used for monitoring staining procedures in pathology laboratories
Envase: Ten unstained slides andMore Product Information