Proteinase K
Supplier: Enzo Life Sciences
ENZ-33801
89165-712EA 87.2 USD
89165-712
Proteinase K
Enzymes
ENZO Proteinase K is specifically designed for use in the pretreatment of formalin-fixed tissue and paraformaldehyde-fixed cells in preparation for in situ hybridization and detection studies.