Anisomycin ≥97% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate

BML-ST102-0050
89164-784EA 276.86 USD
89164-784
Anisomycin ≥97% (by TLC)
Anisomycin

Activator of MAPK, Jnk and p38


Isolated from Streptomyces griseolus.


Antibiotic. Activates JNK/SAPKs, p38 and MAP kinases but not ERKs leading to the phosphorylation of S6, histone H3 and HMG-14 and the induction of the immediate-early genes c-fos and c-jun. Induces apoptosis in the human monoblastoid cell line. Used at 300 ng/ml. Inhibits peptidyl transferase at higher concentrations. Used in the eradication of bean mildew. Inhibits other pathogenic fungi in plants. 

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD00077650
CAS Number: 22862-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR