Cam Kinase II
Supplier: Enzo Life Sciences
BML-P200-0500
89161-064EA 197.29 USD
89161-064
Cam Kinase II
Proteins and Peptides
CaM kinase inhibitor
Synthetic peptide with sequence from Ca2+/calmodulin-dependent protein kinase II pseudosubstrate site. Peptide binds calmodulin tightly and inhibits calmodulin-requiring enzymes including CaMKII, phosphodiesterase and calcineurin. IC50=52 nM for CaMKII in the presence of 100 nM calmodulin.