Cathepsin S Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK431-0001
89156-494EA 709.59 USD
89156-494
Cathepsin S Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZyme™ Assay System
  • Complete system designed to screen modulators of cathepsin S activity
  • The assays are performed in a convenient 96-well microplate format
The Cathepsin S Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin S. Using a convenient microplate format, cathepsin S activity is monitored fluorimetrically using the substrate provided in the kit.
Información de suministro: Kit contains Cathepsin S enzyme (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer and 1/2-volume white microplate.