SCREEN-WELL® Histaminergic Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2816-0500 BML-2816-0100
89156-278EA 1517.28 USD
89156-278 76002-334
SCREEN-WELL® Histaminergic Ligand Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Histaminergic and Melatonin Ligands Plate contains 41 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Histaminergic and Melatonin Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 680 ligands.