5-AIQ hydrochloride ≥97% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 5-Aminoisoquinolinone hydrochloride

270-285-M005
89147-560EA 80.7 USD
89147-560 89147-562
5-AIQ hydrochloride ≥97% (by HPLC)
5-AIQ hydrochloride

Water-soluble, potent inhibitor of poly(ADP-ribose) polymerase-1 (PARP-1).


  • PARP-1 inhibitor


Beige to brown powder.


Soluble in water (warm).

Formula: C₉H₈N₂O·HCl
MW: 160.2:36.5 g/mol
Storage Temperature: Refrigerator
CAS Number: 93117-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feb549aa6b8d6d12451eeeddf0f23e801eef19a9&vid=cc4032a32c56330aa7c9c1040fadd5d5f71a3ea2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR