Comet SCGE Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-166
89141-246EA 420.74 USD
89141-246
Comet SCGE Assay Kit, Enzo Life Sciences
Assays Cellular Assays
COMET SCGE Assay measures DNA damage by fluorescently detecting the integrity of DNA liberated from cells embedded in low melting point agarose. Upon electrophoresis, fragmented DNA produces a characteristic "comet" shaped tail as small DNA fragments migrate in the gel more rapidly than in-tact genomic DNA.
  • Comet Slides allow direct application of sample without pretreatment
  • Higher throughput sample analysis
  • Hydrophobic barrier allows sample treatment with DNA repair enzymes
For the measurement of DNA Damage, DNA Repair, and Apoptosis in cells and tissue from any species via electrophoresis.
The COMET SCGE Assay is a fast and simple electrophoresis method to detect and quantitate DNA fragmentation in cells associated with DNA damage and apoptosis. A unique nucleic acid stain provides improved sensitivity for DNA visualization compared to ethidium bromide.
Información de suministro: Kit includes lysis solution, comet LM agarose, comet slide, EDTA, CYGREEN® nucleic acid dye.