SOD Activity Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-157
89141-228EA 849.11 USD
89141-228
SOD Activity Kit, Enzo Life Sciences
Assays Protein Assays
ELS offers a large collection of superoxide dismutase products for oxidative stress research.
For the measurement of total SOD activity in cell lysates and tissue from any species.
This is a versatile assay for the determination of SOD activity. Differentiate between the activity of three SOD isoenzymes: MnSOD, FeSOD, and Cu/ZnSOD. Obtain higher throughput sample analysis with a convenient colorimetric 96 well plate format and fast results in just 10 minutes.
Información para pedidos: Contents: Microtiter plate, Standard, SOD buffer concentrate, Xanthine oxidase, Xanthine solution, Triton x-100, WST-1 reagent