7-Nitro-1H-indazole

Supplier: Biotium

Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole

00240
89138-166EA 152.85 USD
89138-166
7-Nitro-1H-indazole
7-Nitro-1H-indazole
Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 190 °C
Density: 1.56 g/cm³ (293 °C)
MDL Number: MFCD00022789
CAS Number: 2942-42-9
EINECS: 220-934-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR