Staurosporine

Supplier: Biotium

Synonyms: (5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282

00025
89138-114EA 136.48 USD
89138-114
Staurosporine
Staurosporine

Staurosporine is a potent apoptosis inducer.


  • Potent and cell-permeable inhibitor
  • Induces apoptosis in human neuroblastoma cells
  • Apoptosis in chick embryonic neurons


Staurosporine is a potent and cell-permeable inhibitor of a wide variety of protein kinases including protein kinase C (IC50 = 0.7 nM), protein kinase A (IC50 = 7 nM), and protein kinase G (IC50 = 8.5 nM). Staurosporine induces apoptosis in human neuroblastoma cells and chick embryonic neurons.

Formula: C₂₈H₂₆N₄O₃
MW: 466.541 g/mol
MDL Number: MFCD18252446
CAS Number: 62996-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR