Lab-Tips® Long Knitted Polyester Swab, Berkshire
Supplier: Berkshire
LTP1465.10
89137-172CS 480.61 USD
89137-172
Lab-Tips® Long Knitted Polyester Swab, Berkshire
Swabs Swabs for Cleaning
Berkshire's Lab-Tips® swabs are exceptionally pure for precision cleaning, surface sampling, applying lubricants and other liquids, and validation procedures.
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, acetone, and other solvents
  • High absorbency
  • Lightweight, high absorbency material design reduces actual cost in use compared to heavier weight products
Berkshire's knitted polyester swabs are made from clean room laundered, continuous filament polyester for exceptional purity. TheyMore Product Information