BRAND PCR 8 Strips Tubes, BrandTech®
Supplier: Brandtech
781325
89135-432PK 299.94 USD
89135-432
BRAND PCR 8 Strips Tubes, BrandTech®
Tubes PCR Tubes
Tubes with flat caps are suitable for Real Time PCR (qPCR)
  • Easy to open and close without tools
  • The tubes have a holding strap at one end, and the cover strips have a small lip on the side of each cap for careful, contamination-free opening
  • DNase, DNA and RNase-free
  • Autoclavable 121 °C (20 min)
8 connected 0.2 ml tubes for separate flat caps in strips of 8.