Organic Vapor Filter , Quality Lab Accessories
Supplier: QLA
EF-101
89132-996EA 98 USD
89132-996
Organic Vapor Filter , Quality Lab Accessories
Filters Venting Filters
Organic vapor filter.
Used on SLVSAF-01 and SLVSAF-02 solvent safe HPLC waste collection products.