Digital Depth Gage Assembly, Quality Lab Accessories
Supplier: QLA
DDAPT-Q
89132-992EA 680.59 USD
89132-992
Digital Depth Gage Assembly, Quality Lab Accessories
Tool Sets
Digital Depth Gage Assembly for accurate measurement of Dissolution Paddle and Basket height within the dissolution vessel.
Información de suministro: Includes all required components and storage case.