Azure A Staining (Gram)

Supplier: VWR International

Synonyms: Azurol a, N,N-Dimethylthionine, Azure A chloride, azurol I, 7 - (dimethyl) - 3-imino-3h-dithizine hydrochloride, 3-amino-7-dimethylaminophenothiazine-5-onium chloride, 3-Amino-7-(dimethylamino)phenothiazin-5-ium chloride, 3-Amino-7-(dimethylamino)phenazathionium Chloride, Methylene Azur A

89130-184
89130-184EA 280.83 USD
89130-184
Azure A Staining (Gram)
Azure A

Used in histological staining of tissues.


  • Used on blood smears.


Azure A is used making azure eosin stains for blood smears.

Formula: C₁₄H₁₄ClN₃S
MW: 291.797 g/mol
MDL Number: MFCD00012112
CAS Number: 531-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR