HiPrep Q XL 16/10 Columns, Cytiva
Supplier: Cytiva
28-9365-38
89128-884EA 983.1 USD
89128-884
HiPrep Q XL 16/10 Columns, Cytiva
Chromatography Columns
HiPrep SP XL 16/10 is a prepacked, ready to use column for ion exchange chromatography.
  • Provide high loading capacity and fast purifications
  • Optimized for reliable, reproducible separations
  • Compatible with simple pump-based configurations and chromatography systems such as ÄKTA design
  • Excellent choice for rapid enrichment during initial capture of proteins from start material
  • Strong quaternary ammonium (Q) anion exchanger
It provides fast, preparative separations of proteins and other biomolecules.