DuPont™ Tyvek® IsoClean® Boot Covers with PVC Sole
Supplier: DuPont
IC457SWHLG01000B IC457SWHMD01000B IC457SWHXL01000S IC457SWHSM01000S IC457SWHXL01000B IC457SWHSM01000B IC457SWHLG01000S IC457SWHMD01000S
89125-636CS 404.81 USD
89125-636 89127-326 89127-332 89125-638 89127-330 89128-796 89127-324 89127-328
DuPont™ Tyvek® IsoClean® Boot Covers with PVC Sole
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • PVC sole
  • Elastic opening
  • Ankle ties
  • Coverage up to calf
  • Serged seams
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Ankle ties allow for a secure fit. PVC soles provide enhanced skid-resistance and durability.