VWR® TraceClean® LDPE Cubitainers (Collapsible Containers), Natural
Supplier: VWR International
89126-634 89126-636
89126-634CS 292.4 USD
89126-634 89126-636
VWR® TraceClean® LDPE Cubitainers (Collapsible Containers), Natural
Bottles Water Sampling Bottles
These TraceClean® containers are cleaned by Protocol “A”, and are used primarily for grab and compositing of aqueous samples for testing metals and inorganic analytes.
  • LDPE construction
  • Collapsible design
  • Good impact strength
  • Resistant to stress cracks and chemicals
Primary uses of the containers cleaned by Protocol 'B' include Grab and compositing of aqueous samples for Bio-Assay studies, and may include field parameters such as conductivity, pH, and dissolved oxygen.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate ofMore Product Information