VWR® TraceClean® Straight Sided Containers, HDPE
Supplier: VWR International
89126-620 89126-622 89126-624
89126-620CS 359.62 USD
89126-620 89126-622 89126-624
VWR® TraceClean® Straight Sided Containers, HDPE
Jars General Use Jars
These TraceClean® containers are used primarily for the collection of soils, clays, slurries, and sediment media for geotechnical testing, Toxicity Characteristic Leaching Procedure (TCLP), metals, total metals, inorganic analytes, and benthic organism studies.
  • HDPE construction
  • Straight-sided design
  • Suited to holding collections of geological materials
Not all analytical parameters are included on the Certificate of Analysis
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analyticalMore Product Information