E/E absolute, KOPTEC, pure (200 Proof)

Supplier: Decon Labs
Danger

Synonyms: Ethyl alcohol

V1005HC V1016 V1016TP V1055TP V1055M V1001 V1005 V1005SG V1055 V1005TP V1016G V1016GTP V1001G V1001GTP V1001TP V1005HCTP V1005SGTP V1055MTP V1005M V1005MTP
71002-512EA 230.57 USD
71002-512 89125-170 89125-186 89125-192 71001-624 89125-172 89125-174 71001-628 89125-176 89125-190 71001-862 71001-866 71002-422 71002-426 89125-188 89234-848 89234-850 89234-852 89426-252 89501-404
E/E absolute, KOPTEC, pure (200 Proof)
Ethanol

Pure (undenatured) anhydrous (200 proof/100%) ethyl alcohol meets USP and Multicompendial specifications used as a cleaner, solvent, or reagent.


  • Total alcohol: 99.5%
  • Ethyl alcohol: 99.5%
  • Denaturant: none


Used in research, healthcare, industrial, and production. 200 proof ethanol is commonly referred to as "anhydrous." Koptec's water specification is less than 0.1%.


Información para pedidos: Tax-paid ethanol prices include the federal excise tax on pure ethanol. To purchase tax-free ethanol, customers must possess an Industrial Alcohol User Permit from TTB specifically marked for the withdrawal and use of alcohol free of tax.

Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78.3
Melting Pt: -117
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Max. UV Absorbance at 240 nm 0.40 AU
Max. UV Absorbance at 250 nm 0.3 AU
Max. UV Absorbance at 260 nm 0.3 AU
Max. UV Absorbance at 270 nm 0.1
Acidity 30 ppm max.
Alkalinity Pass test
Assay 200 proof (99.5% min.)
Benzene 2 ppm max.
Clarity Pass
Color Pass test
Methanol 100 ppm max.
Non-Volatile Residue 25 ppm max.
Relative Density at 20°C (BP/EP) 0.790–0.793
Specific Gravity at 15.0°C (JP) 0.797 max.
Specific Gravity at 15.56°C (USP) 0.7962 max.
Sum of Acetal & Acetaldehyde 10 ppm max.
Water 0.1% (v/v) max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR