EVA PAPRs (Powered Air Purifying Respirators), Complete Systems, Bullard®
Supplier: Bullard
EVA20LF2L EVA20TJ EVA20TIC EVA20TICH EVART3
89097-702EA 2284.93 USD
89097-702 89097-704 89097-706 89097-710 89097-714 89097-718 89097-722
EVA PAPRs (Powered Air Purifying Respirators), Complete Systems, Bullard®
Respirators Respiratory Systems
The EVA is simple, safe, and dependable.
  • Versatile respirator for particulate or chemical environments
  • Lightweight, compact, well-balanced design
  • Compatible with range of head tops
  • Standard comfort belt
In addition, the EVA offers Active Flow Technology™, which maintains an airflow of 8.5 cfm (high speed), 1000 Assigned Protection Factor (depending upon head top), and dual alarms for low flow and low battery. Finally, the EVA has a visual battery fuel gauge to check remaining charge at the push of a button, a brushless motor with ten times the life ofMore Product Information