VWR® Traceable® Jumbo Refrigerator/Freezer Thermometers
Supplier: VWR International
89094-770 89094-772 89094-768
89094-770EA 115.31 USD
89094-770 89094-772 89094-768
VWR® Traceable® Jumbo Refrigerator/Freezer Thermometers
Thermometers Digital Thermometers
Perfect for monitoring freezers, heating blocks, incubators, refrigerators, and water baths.
  • Traceable
  • Temperature in °C and °F
  • Simultaneously displays minimum/maximum temperatures
  • Solution insulates against rapid temperature changes
  • Alarm feature for significant temperature changes
Solid-state sensor probe eliminates mercury contamination
Thermometer 89094-770 has a sensor probe that is sealed in a 2.5x6.4 cm (1x21/2") bottle filled with GRAS nontoxic glycol. The solution insulates the sensor from rapid temperature changes, such as whenMore Product Information