Dura Flock® Flock-Lined Nitrile Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
DFK-608-S DFK-608-L DFK-608-XL DFK-608-M
89094-568CS 600.02 USD
89094-568 89094-572 89094-574 89094-576 89094-570
Dura Flock® Flock-Lined Nitrile Gloves, Microflex®, Ansell
Gloves
Ambidextrous, powder-free, 100% nitrile gloves feature an ultra-thin absorbent flock lining to keep hands dry, comfortable, and protected.
  • Beaded cuffs
  • Fully textured for enhanced grip
  • Comfortable grip for reduced hand fatigue
8-millimeter gloves are thicker than standard disposable gloves for increased durability and superior protection.