VWR® TraceClean® Cubitainer Containers, LDPE
Supplier: VWR International
89094-136 89094-138 89094-140 89094-142 89094-144
89094-136CS 194.62 USD
89094-136 89094-138 89094-140 89094-142 89094-144
VWR® TraceClean® Cubitainer Containers, LDPE
Bottles Water Sampling Bottles
Containers are for collecting groundwater and influent, effluent, and bio-assay waters.
  • Low-density polyethylene construction
  • Completely traceable
  • Custody-sealed to ensure content integrity
  • Supplied with 38-400 polyfoam-lined caps
Processed (PC) and Quality-Assured® (QA) containers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'.
QA containers include a Certificate of Analysis in every case, as well as bar codes with lot number and individual containerMore Product Information