VWR® TraceClean® Straight-Sided Wide Mouth Jars, Glass
Supplier: VWR International
89093-938 89093-940 89093-946 89093-948 89093-954 89093-956 89093-968 89093-970 89093-972 89093-974 89093-976 89093-978 89093-980 89093-982 89093-984 89093-986 89093-988 89093-990 89094-016 89094-024 89094-028 89094-030 89094-032 89094-018
89093-938CS 169.08 USD
89093-938 89093-940 89093-946 89093-948 89093-954 89093-956 89093-968 89093-970 89093-972 89093-974 89093-976 89093-978 89093-980 89093-982 89093-984 89093-986 89093-988 89093-990 89094-016 89094-024 89094-028 89094-030 89094-032 89094-018
VWR® TraceClean® Straight-Sided Wide Mouth Jars, Glass
Jars General Use Jars
Type III (soda-lime) glass jars are for collection of soil, sediment, and slurry matrices.
  • Amber glass fabrication
  • Straight sides
  • Wide-mouthed design
  • Recommended for UV light-sensitive samples
  • Jars are available with solid-top or open-top caps
Processed (PC) and Quality-Assured® (QA) jars follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA jars include a Certificate of Analysis in every case.
QA jars also include bar codes with lot number and individualMore Product Information