VWR® TraceClean® Wide Mouth Jars, Glass
Supplier: VWR International
89093-942 89093-944 89093-950 89093-958 89093-960 89093-992 89093-994 89093-996 89093-998 89094-000 89094-002 89094-004 89094-006 89094-008 89094-010 89094-012 89094-014 89093-952
89093-942CS 161.49 USD
89093-942 89093-944 89093-950 89093-958 89093-960 89093-992 89093-994 89093-996 89093-998 89094-000 89094-002 89094-004 89094-006 89094-008 89094-010 89094-012 89094-014 89093-952
VWR® TraceClean® Wide Mouth Jars, Glass
Jars General Use Jars
Type III jars are for collection of solid matrices or liquids dispensed from bailers or peristaltic tubing. Jars are available with solid-top or open-top caps. Solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, and PCBs; and select inorganic compounds. Open-top caps with ultrasonically bonded fluoropolymer resin/silicone septa are for use in volatile organic analysis.
  • Soda-lime glass fabrication
  • Wide-mouthed design
  • Available with solid-top or open-top caps
  • Individually numbered for traceability
  • Jars are available with
More Product Information