VWR® TraceClean® Boston Round Bottles, Glass
Supplier: VWR International
89093-878 89093-880 89093-886 89093-888 89093-890 89093-892 89093-894 89093-900 89093-902 89093-904 89093-906 89093-908 89093-914 89093-916 89093-918 89093-920 89093-922 89093-924 89093-926 89093-928 89093-932 89093-934 89093-936 89095-160 89095-162 89096-742 89096-744 89096-746 89093-882 89093-884 89093-896 89093-898 89093-910 89093-930 89093-912
89093-878CS 64.7 USD
89093-878 89093-880 89093-886 89093-888 89093-890 89093-892 89093-894 89093-900 89093-902 89093-904 89093-906 89093-908 89093-914 89093-916 89093-918 89093-920 89093-922 89093-924 89093-926 89093-928 89093-932 89093-934 89093-936 89095-160 89095-162 89096-742 89096-744 89096-746 89093-882 89093-884 89093-896 89093-898 89093-910 89093-930 89093-912
VWR® TraceClean® Boston Round Bottles, Glass
Bottles Water Sampling Bottles
Soda lime glass bottles are available with solid-top or open-top caps.
  • Soda-lime glass fabrication
  • Available with solid-top or open-top caps
  • Amber glass is ideal for protecting UV light-sensitive compounds
  • Open-top caps feature ultrasonically bonded fluoropolymer resin/silicone septum and are for volatile organic analyses
  • Amber glass is recommended for UV light-sensitive compounds
Solid-top caps feature fluoropolymer resin liner and are for semi-volatiles, pesticides, and PCB analyses; and select inorganic compounds.
Processed (PC) and Quality-More Product Information