PYREX® Volumetric Pipettes, Class A, Color-Coded, Graduated, Serialized/Certified, Corning
Supplier: Corning
7101-100 7101-10 7101-1 7101-20 7101-2 7101-4 7101-50 7101-15 7101-25 7101-3 7101-5
89091-082CS 727.61 USD
89091-082 89091-122 89091-226 89091-234 89091-304 89091-392 89091-466 89091-178 89091-270 89091-364 89091-490
PYREX® Volumetric Pipettes, Class A, Color-Coded, Graduated, Serialized/Certified, Corning
Pipettes Graduated Pipettes
Pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
  • Sizes 1ml through 25ml are color-coded for ease in sorting and selection
  • Colored graduations are enameled on to the glass
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Información de suministro: Each pipette is individually serialized and supplied with a certificate of identity and capacity, traceable to NIST standards.