PYREX® Erlenmeyer Flasks, Graduated, [ST] Stopper, Corning
Supplier: Corning
5020-1L 5020-25 5020-50 5020-125 5020-250 5020-500
89091-204CS 547.18 USD
89091-204 89091-266 89091-454 89090-650 89090-788 89090-876
PYREX® Erlenmeyer Flasks, Graduated, [ST] Stopper, Corning
Flasks Erlenmeyer Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth have standard taper stoppers for secure sealing. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance. For convenience, these flasks are graduated to show approximate capacity. The 125 ml flasks are satisfactory for use as weighing bottles.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental Certification I-RECs, ISO 14001 - Monterrey