PYREX® Iodine Determination Flasks, [ST] Stopper, Corning
Supplier: Corning
5400-500 5400-125 5400-250
89091-410CS 620.88 USD
89091-410 89090-640 89090-780
PYREX® Iodine Determination Flasks, [ST] Stopper, Corning
Flasks Iodine Flasks
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Blown in a specially-designed mold to ensure uniformity and proper contour. The stopper handle projects above the liquid seal trough to facilitate removal of the stopper. With No. 22 [ST] stopper. ASTM D29 and D555.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental Certification I-RECs, ISO 14001 - Monterrey