Corning® E-Cube™ Culture System Kit, Corning
Supplier: Corning
3286
89089-916CS 6616.8 USD
89089-916
Corning® E-Cube™ Culture System Kit, Corning
Bioreactors
The E-Cube™ System is a simple bioreactor with 8500 cm² cell growth area for growing anchorage dependent cells in only a 25.4×35.6 cm footprint. The E-Cube™ System provides a simple method to determine if cells will grow in the CellCube® Module prior to investing in the resources and funding that would be necessary for the larger, automated CellCube® System. Peristaltic pump, CO₂ incubator and medium are not provided.
Includes an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings.
Cells grow in Corning's parallel-plate CellCube® Module on the sameMore Product Information