LabKlenz™ 250 Acid Detergents
Supplier: Steris
1L2502 1L2508 1L2505
89089-214EA 1493.96 USD
89089-214 89089-218 89089-216
LabKlenz™ 250 Acid Detergents
Cleaning Agents Detergents and Surface Cleansers
LabKlenz™ 250 Detergent is a low-foaming, citric-acid based liquid detergent.
  • Flexibility - Effective removal of laboratory residues in both soft and hard water
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
LabKlenz™ 250 Detergent is specially formulated for use in automated glassware washing applications both asMore Product Information