Choice® Cleanroom Laundered 100% Polyester Knit Wipe, Berkshire
Supplier: Berkshire
CH709.14
89089-178CS 1038.49 USD
89089-178
Choice® Cleanroom Laundered 100% Polyester Knit Wipe, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ultrasonically sealed edge for reduced fiber contamination.
  • Chemically compatible with IPA, acetone and other solvents
Lightweight, high absorbency design can reduce landfill waste impact when compared to more traditional heavier weight products.
Applications include chamber cleaning and CMP processing, stencil and print roll cleaning, cleaning of medical device products, applying and removing cleaning and disinfecting solutions. Steam autoclavable for aseptic environments.
Certificaciones: Complies with RoHS and REACH requirements.