Axygen® PCR Tube Strips and Caps, Corning
Supplier: Corning
PCR-02CP-C PCR-2CP-RT-C PCR-0208-FCP-C PCR-02-FCP-C PCR-0212-FCP-C PCR-12-FCP-C
10011-786CS 564.33 USD
10011-786 47745-512 89080-524 89080-526 89179-620 89179-624
Axygen® PCR Tube Strips and Caps, Corning
Tubes PCR Tubes
Axygen® polypropylene 8/12-tube thin wall PCR tube strips and caps are specifically designed for PCR use and are ideal for thermal transfer applications. The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling. Made from highly polished moulds and free of lubricants, dyes, heavy metals, and fillers. Certified RNase, DNase, and pyrogen free to ensure the purity of your reaction.
  • Ultra-thin walls of uniform thickness optimise
More Product Information