VWR® Signature™ Ergonomic High Performance Single Channel Variable Volume Pipettors
Supplier: VWR International
P3925-2VWR P3925-10VWR P3925-20VWR P3925-50VWR P3925-100VWR P3925-200VWR P3925-250VWR P3925-1000VWR P3925-5000VWR P3925-10000VWR P3925-SKA3-VWR P3925-SKB3-VWR P3925-SKC4-VWR
89079-960EA 376.28 USD
89079-960 89079-962 89079-964 89079-966 89079-968 89079-970 89079-972 89079-974 89079-976 89079-978 10002-778 10002-780 10002-782 10002-830 10002-834 10002-836 75788-456 75788-458 75788-460 76498-674 76498-676 76498-678
VWR® Signature™ Ergonomic High Performance Single Channel Variable Volume Pipettors
Pipettes Micropipettes
The new, ergonomic design of the pipettors allow for consistently comfortable pipetting even with extended use.
  • Superior accuracy and precision for critical applications
  • Reduced plunger forces
  • Fully autoclavable
  • Volume locking mechanism
An extended, curved finger rest allows the pipettor to balance on the index finger, and both plunger and volume-adjustment forces have been significantly reduced to eliminate the strain of repetitive pipetting.
To accommodate a wide variety of tips, the easy-grip ejector system provides adjustment of the ejectorMore Product Information