Ammonium iron (III) citrate, green, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

F1001-50KGBL F1001-125GM F1001-2.5KG F1001-500GM F1001-12KG
89078-804EA 3039.15 USD
89078-804 95033-208 95033-210 95033-212 75811-110
Ammonium iron (III) citrate, green, powder FCC
Ammonium iron (III) citrate

Ferric Ammonium Citrate, Green, Powder, FCC is also known as Ferric ammonium citrate or Ammonium Iron(III) Citrate and is used as an acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5

Order Now

Specification Test Results

Assay (Fe) 14.5 - 16.0 %
Ferric Citrate To Pass Test
Oxalate (C₂O₄) To Pass Test
Sulfate max. 0.3 %
Mercury max. 1 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR