PT100 Temperature Sensor, Ace Glass Incorporated
Supplier: Ace Glass
12103-18 12103-19 12103-20 12103-21
89076-408EA 429.42 USD
89076-408 89076-410 89076-412 89076-414
PT100 Temperature Sensor, Ace Glass Incorporated
Temperature Probes
The PT100 (100 ohm platinum wire resistance temperature detector) probes provide measurement over a temperature range of -20°C to 550°C
Supplied with male phone plug ends.